Learn More
1-Fluoro-2,4-dinitrobenzene, 99%, Thermo Scientific Chemicals
Description
1-Fluoro-2,4-dinitrobenzene is used to identify the amino acid sequence. It reacts with amino group of amino acids to yield dinitrophenyl-amino acids. It is also used in chromatographic methods. Further, it acts as an alkylating agent used in elucidating amino acid sequence in proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Fluoro-2,4-dinitrobenzene is used to identify the amino acid sequence. It reacts with amino group of amino acids to yield dinitrophenyl-amino acids. It is also used in chromatographic methods. Further, it acts as an alkylating agent used in elucidating amino acid sequence in proteins.
Solubility
Soluble in chloroform, benzene, ether and propylene glycol.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
Specifications
Melting Point | 24°C to 30°C |
Density | 1.482 |
Boiling Point | 296°C |
Flash Point | >110°C (230°F) |
Refractive Index | 1.569 |
Quantity | 100 g |
UN Number | UN2811 |
Beilstein | 398632 |
Merck Index | 14,4172 |
Solubility Information | Soluble in chloroform,benzene,ether and propylene glycol. |
Show More |
Safety and Handling
Hazard Category | H301+H311+H331-H315-H317-H320-H335-H336-H373 |
Hazard Statement | GHS H Statement H301-H311-H331-H318-H341-H315-H317 Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes serious eye damage. Suspected of causing genetic defects. Causes skin irritation. May cause an allergic skin reaction. |
Precautionary Statement | P260-P262-P264b-P270-P271-P272-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P314-P330-P333+P313-P362-P501c |
DOTInformation | Hazard Class: 6.1; Packaging Group: II |
EINECSNumber | 200-734-3 |
RTECSNumber | CZ7800000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.