Alfa Aesar™ 1-Fluoro-2,4-dinitrobenzene, 99%

Manufacturer: Alfa Aesar™ A1187106

Quantity 5g

 View more versions of this product

Catalog No. AAA1187106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 70-34-8
    Molecular Formula C6H3FN2O4
    Formula Weight 186.1
    MDL Number MFCD00007056
    UN Number UN2811
    Beilstein 398632
    Merck Index 14,4172
    Synonym 2,4-Dinitrofluorobenzene; Sanger's Reagent
    Chemical Name or Material 1-Fluoro-2,4-dinitrobenzene

    Specifications

    Formula Weight 186.1
    Health Hazard 2 GHS H Statement
    H301-H311-H331-H318-H341-H315-H317
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes serious eye damage.
    Suspected of causing genetic defects.
    Causes skin irritation.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 70-34-8
    UN Number UN2811
    RTECS Number CZ7800000
    Flash Point >110°C (230°F)
    Boiling Point 296°C
    TSCA TSCA
    Molecular Formula C6H3FN2O4
    MDL Number MFCD00007056
    Beilstein 398632
    Melting Point 24° to 30°C
    Quantity 5g
    Synonym 2,4-Dinitrofluorobenzene; Sanger's Reagent
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Merck Index 14,4172
    Chemical Name or Material 1-Fluoro-2,4-dinitrobenzene
    Assay 99%

    Description

    Applications
    1-Fluoro-2,4-dinitrobenzene is used to identify the amino acid sequence. It reacts with amino group of amino acids to yield dinitrophenyl-amino acids. It is also used in chromatographic methods. Further, it acts as an alkylating agent used in elucidating amino acid sequence in proteins.

    Solubility
    Soluble in chloroform, benzene, ether and propylene glycol.

    Notes
    Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong bases.

    GHS H Statement
    H301-H311-H331-H318-H341-H315-H317
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes serious eye damage.
    Suspected of causing genetic defects.
    Causes skin irritation.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit