Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Fluoro-2,6-dichloropyridinium Tetrafluoroborate 96.0+%, TCI America™
Click to view available options
Quantity:
5g
25g
Specifications
Melting Point | 245°C |
Color | Yellow |
Quantity | 5g |
UN Number | 1759 |
Formula Weight | 253.79 |
Physical Form | Crystal-Powder at 20°C |
Percent Purity | ≥96.0% (T) |
Chemical Name or Material | 1-Fluoro-2,6-dichloropyridinium Tetrafluoroborate [Fluorinating Reagent] |
Safety and Handling
TSCA : No