Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Fluoro-2-nitrobenzene, 99%, ACROS Organics™

Click to view available options
Quantity:
10mL
50mL
250mL
Packaging:
Glass bottle
1493-27-2
C6H4FNO2
141.1
MFCD00007048
PWKNBLFSJAVFAB-UHFFFAOYSA-N
2-fluoronitrobenzene,o-fluoronitrobenzene,o-nitrofluorobenzene,2-nitrofluorobenzene,benzene, 1-fluoro-2-nitro,benzene, o-nitrofluoro,2-fluoro-1-nitrobenzene,fluoronitrobenzene,1-fluoro-2-nitro-benzene,benzene, fluoronitro
73895
1-fluoro-2-nitrobenzene
C1=CC=C(C(=C1)[N+](=O)[O-])F
1493-27-2
C6H4FNO2
141.1
MFCD00007048
PWKNBLFSJAVFAB-UHFFFAOYSA-N
2-fluoronitrobenzene,o-fluoronitrobenzene,o-nitrofluorobenzene,2-nitrofluorobenzene,benzene, 1-fluoro-2-nitro,benzene, o-nitrofluoro,2-fluoro-1-nitrobenzene,fluoronitrobenzene,1-fluoro-2-nitro-benzene,benzene, fluoronitro
73895
1-fluoro-2-nitrobenzene
C1=CC=C(C(=C1)[N+](=O)[O-])F
Melting Point -9°C to 6°C
Boiling Point 215°C
Flash Point 94°C
Specific Gravity 1.335
Refractive Index 1.5307 to 1.5327
Density 1.3350g/mL
Quantity 250mL
Packaging Glass bottle
Color Brown to Yellow
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula FC6H4NO2
Beilstein 05, 241
Solubility Information Solubility in water: immiscible
Formula Weight 141.1
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 1-Fluoro-2-nitrobenzene
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning