Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Fluoro-3,5-dimethoxybenzene, 98%

Click to view available options
Quantity:
5g
25g
100g
52189-63-6
C8H9FO2
156.156
MFCD00012445
IWFKMNAEFPEIOY-UHFFFAOYSA-N
1,3-dimethoxy-5-fluorobenzene,benzene, 1-fluoro-3,5-dimethoxy,3,5-dimethoxyfluorobenzene,pubchem3049,acmc-209wz0,3,5-dimethoxy fluorobenzene,ksc595k6b,3,5-dimethoxy-1-fluorobenzene,1-fluoro-3,5-dimethoxy benzene,benzene,1-fluoro-3,5-dimethoxy
2774257
1-fluoro-3,5-dimethoxybenzene
COC1=CC(=CC(=C1)F)OC
52189-63-6
C8H9FO2
156.156
MFCD00012445
IWFKMNAEFPEIOY-UHFFFAOYSA-N
1,3-dimethoxy-5-fluorobenzene,benzene, 1-fluoro-3,5-dimethoxy,3,5-dimethoxyfluorobenzene,pubchem3049,acmc-209wz0,3,5-dimethoxy fluorobenzene,ksc595k6b,3,5-dimethoxy-1-fluorobenzene,1-fluoro-3,5-dimethoxy benzene,benzene,1-fluoro-3,5-dimethoxy
2774257
1-fluoro-3,5-dimethoxybenzene
COC1=CC(=CC(=C1)F)OC
Applications
1-Fluoro-3,5-dimethoxybenzene is used in the preparation of 5-fluoro-benzene-1,3-diol using reagents like tetrabutylammnoium iodide and boron trichloirde.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
Quantity 100g
Boiling Point 86°C to 88°C (7mmHg)
Flash Point 91°C (196°F)
Formula Weight 156.16
Percent Purity 98%
Chemical Name or Material 1-Fluoro-3,5-dimethoxybenzene

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning