Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Fluoro-3-nitrobenzene, 98+%

Click to view available options
Quantity:
25g
100g
402-67-5
C6H4FNO2
141.101
MFCD00007196
WMASLRCNNKMRFP-UHFFFAOYSA-N
3-fluoronitrobenzene,m-fluoronitrobenzene,m-nitrofluorobenzene,benzene, 1-fluoro-3-nitro,3-fluoro-1-nitrobenzene,1-fluoro-3-nitro-benzene,3-fluoro-nitrobenzene,3-nitro-fluorobenzene,pubchem2286,3-fluoro nitro benzene
9823
1-fluoro-3-nitrobenzene
C1=CC(=CC(=C1)F)[N+](=O)[O-]
402-67-5
C6H4FNO2
141.101
MFCD00007196
WMASLRCNNKMRFP-UHFFFAOYSA-N
3-fluoronitrobenzene,m-fluoronitrobenzene,m-nitrofluorobenzene,benzene, 1-fluoro-3-nitro,3-fluoro-1-nitrobenzene,1-fluoro-3-nitro-benzene,3-fluoro-nitrobenzene,3-nitro-fluorobenzene,pubchem2286,3-fluoro nitro benzene
9823
1-fluoro-3-nitrobenzene
C1=CC(=CC(=C1)F)[N+](=O)[O-]
Melting Point 2°C
Boiling Point 204°C to 206°C
Flash Point 76°C (168°F)
Quantity 25g
UN Number UN2810
Beilstein 1862210
Formula Weight 141.1
Percent Purity ≥98%
Chemical Name or Material 1-Fluoro-3-nitrobenzene

GHS H Statement
H301-H311-H331-H373-H227-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P210-P301+P310-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

RTECSNumber : DA1385000