Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 1-Fluoro-3-nitrobenzene, 98+%
Click to view available options
Quantity:
25g
100g
Specifications
Refractive Index | 1.525 |
Density | 1.325 |
Melting Point | 2°C |
Flash Point | 76°C (168°F) |
Boiling Point | 204°C to 206°C |
Quantity | 100g |
UN Number | UN2810 |
Beilstein | 1862210 |
Formula Weight | 141.1 |
Percent Purity | ≥98% |
Show More |
Safety and Handling
GHS H Statement
H301-H311-H331-H373-H227-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
P210-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P370+P378q-P501c
H227-H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 206-953-0
RTECSNumber : DA1385000
TSCA : No
Recommended Storage : Ambient temperatures