Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Fluoro-4-iodobenzene, 99%, ACROS Organics™

Click to view available options
Quantity:
1mL
5mL
50mL
Packaging:
Glass bottle
352-34-1
C6H4FI
222
MFCD00001052
KGNQDBQYEBMPFZ-UHFFFAOYSA-N
4-fluoroiodobenzene,p-fluoroiodobenzene,benzene, 1-fluoro-4-iodo,4-iodofluorobenzene,p-iodofluorobenzene,4-fluoro-1-iodobenzene,4-fluoro-iodobenzene,1-fluoro-4-iodo-benzene,1-iodo-4-fluorobenzene,4-fluroiodobenzene
9605
1-fluoro-4-iodobenzene
C1=CC(=CC=C1F)I
352-34-1
C6H4FI
222
MFCD00001052
KGNQDBQYEBMPFZ-UHFFFAOYSA-N
4-fluoroiodobenzene,p-fluoroiodobenzene,benzene, 1-fluoro-4-iodo,4-iodofluorobenzene,p-iodofluorobenzene,4-fluoro-1-iodobenzene,4-fluoro-iodobenzene,1-fluoro-4-iodo-benzene,1-iodo-4-fluorobenzene,4-fluroiodobenzene
9605
1-fluoro-4-iodobenzene
C1=CC(=CC=C1F)I
Specific Gravity 1.925
Boiling Point 182°C to 184°C
Flash Point 68°C
Melting Point -20°C
Packaging Glass bottle
Density 1.9250g/mL
Refractive Index 1.5820 to 1.584
Color Yellow
Infrared Spectrum Authentic
Quantity 5mL
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula FC6H4I
Beilstein 05, 220
Formula Weight 222
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 1-Fluoro-4-iodobenzene
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 206-522-7

Recommended Storage : Product darkens on storage