Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Fluoro-4-nitrobenzene, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
350-46-9
C6H4FNO2
141.1
MFCD00007282
WFQDTOYDVUWQMS-UHFFFAOYSA-N
4-fluoronitrobenzene,p-fluoronitrobenzene,p-nitrofluorobenzene,benzene, 1-fluoro-4-nitro,4-nitrofluorobenzene,1-nitro-4-fluorobenzene,4-fluoro-1-nitrobenzene,1-fluoro-4-nitro-benzene,unii-a2m2fh7xhh,p-fluoro nitrobenzene
9590
1-fluoro-4-nitrobenzene
C1=CC(=CC=C1[N+](=O)[O-])F
350-46-9
C6H4FNO2
141.1
MFCD00007282
WFQDTOYDVUWQMS-UHFFFAOYSA-N
4-fluoronitrobenzene,p-fluoronitrobenzene,p-nitrofluorobenzene,benzene, 1-fluoro-4-nitro,4-nitrofluorobenzene,1-nitro-4-fluorobenzene,4-fluoro-1-nitrobenzene,1-fluoro-4-nitro-benzene,unii-a2m2fh7xhh,p-fluoro nitrobenzene
9590
1-fluoro-4-nitrobenzene
C1=CC(=CC=C1[N+](=O)[O-])F
Melting Point 27°C
Flash Point 83°C
Specific Gravity 1.33
Boiling Point 205°C
Density 1.3300g/mL
Infrared Spectrum Authentic
Refractive Index 1.5302 to 1.5322
Color Brown to Yellow
Quantity 500g
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula FC6H4NO2
Beilstein 05, 241
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,xylene and acetone
Formula Weight 141.1
Physical Form Liquid After Melting
Percent Purity 99%
Chemical Name or Material 1-Fluoro-4-nitrobenzene, 99%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
M

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dus

GHS Signal Word: Warning

EINECSNumber : 206-502-8

RTECSNumber : DA1400000

TSCA : TSCA