Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Formylpiperidine, 99%

Click to view available options
Quantity:
50g
250g
1000g
2591-86-8
C6H11NO
113.16
MFCD00006483
FEWLNYSYJNLUOO-UHFFFAOYSA-N
n-formylpiperidine,1-formylpiperidine,1-piperidinecarboxaldehyde,formylpiperidine,n-formylpiperidin,piperidinoformamide,piperidine-n-carbaldehyde,piperidine, 1-formyl,1-piperidinecarbaldehyde,unii-ziq29h6czg
17429
CHEBI:42546
piperidine-1-carbaldehyde
C1CCN(CC1)C=O
2591-86-8
C6H11NO
113.16
MFCD00006483
FEWLNYSYJNLUOO-UHFFFAOYSA-N
n-formylpiperidine,1-formylpiperidine,1-piperidinecarboxaldehyde,formylpiperidine,n-formylpiperidin,piperidinoformamide,piperidine-n-carbaldehyde,piperidine, 1-formyl,1-piperidinecarbaldehyde,unii-ziq29h6czg
17429
CHEBI:42546
piperidine-1-carbaldehyde
C1CCN(CC1)C=O
Applications
1-Formylpiperidine reacts with Grignard or alkyllithium reagents gives aldehydes in high yield. It can be used as a polar aprotic solvent, with better hydrocarbon solubility than other amide solvents such as dimethylformamide. In some formylation reaction of alkyllithium compounds it gives higher yields than the DMF.

Solubility
Freely soluble in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Quantity 250g
Flash Point 102°C (215°F)
Melting Point -31°C
Boiling Point 222°C
Beilstein 107697
Formula Weight 113.16
Percent Purity 99%
Chemical Name or Material 1-Formylpiperidine

GHS H Statement
H301-H311-H227-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P301+P310-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : TN0380000

TSCA : TSCA