Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Furfurylpyrrole, 99%

Manufacturer:  Alfa Aesar™ B2310506

Quantity 5g

 View more versions of this product

Catalog No. AAB2310506


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1438-94-4
147.18
UN2810
1-Furfurylpyrrole
C9H9NO
MFCD00010080
1-(2-Furylmethyl)-1H-pyrrole

Specifications

1438-94-4
147.18
UN2810
76° to 78°C (1mmHg)
92°C (199°F)
99%
C9H9NO
MFCD00010080
1-(2-Furylmethyl)-1H-pyrrole
5g
1-Furfurylpyrrole

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.

GHS P Statement
P210-P280-P264-P301+P312-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

RTECSNumber : UX9631000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit