Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Furfurylpyrrole, 99%

Click to view available options
Quantity:
5g
25g
100g
1438-94-4
C9H9NO
147.177
MFCD00010080
BTBFUBUCCJKJOZ-UHFFFAOYSA-N
1-furfurylpyrrole,n-furfurylpyrrole,1-furan-2-ylmethyl-1h-pyrrole,1-furfuryl-1h-pyrrole,1h-pyrrole, 1-2-furanylmethyl,1-2-furanylmethyl-1h-pyrrole,n-2-furfuryl pyrrole,n-furfuryl pyrrole,pyrrole, 1-furfuryl,1-2-furfuryl pyrrole
15037
1-(furan-2-ylmethyl)pyrrole
C1=CN(C=C1)CC2=CC=CO2
1438-94-4
C9H9NO
147.177
MFCD00010080
BTBFUBUCCJKJOZ-UHFFFAOYSA-N
1-furfurylpyrrole,n-furfurylpyrrole,1-furan-2-ylmethyl-1h-pyrrole,1-furfuryl-1h-pyrrole,1h-pyrrole, 1-2-furanylmethyl,1-2-furanylmethyl-1h-pyrrole,n-2-furfuryl pyrrole,n-furfuryl pyrrole,pyrrole, 1-furfuryl,1-2-furfuryl pyrrole
15037
1-(furan-2-ylmethyl)pyrrole
C1=CN(C=C1)CC2=CC=CO2
Density 1.081
Refractive Index 1.531
Sensitivity Light sensitive
Boiling Point 76°C to 78°C (1mmHg)
Quantity 100g
Flash Point 92°C (199°F)
UN Number UN2810
Formula Weight 147.18
Percent Purity 99%
Chemical Name or Material 1-Furfurylpyrrole

GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.

P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c

H227-H302

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 215-876-1

RTECSNumber : UX9631000

TSCA : Yes

Recommended Storage : Ambient temperatures