Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Heptanesulfonic acid, sodium salt, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Glass bottle
22767-50-6
C7H15NaO3S
202.25
REFMEZARFCPESH-UHFFFAOYSA-M
sodium 1-heptanesulfonate,sodium heptane-1-sulfonate,1-heptanesulfonic acid sodium salt,1-heptanesulfonic acid, sodium salt,sodium heptane-1-sulphonate,1-heptanesulfonic acid, sodium salt 1:1,1-heptanesulfonic acid sodium,sodium heptanesulfonate,1-heptylsodiumsulfonate,sodium heptane sulfonate
23672332
sodium;heptane-1-sulfonate
CCCCCCCS(=O)(=O)[O-].[Na+]
22767-50-6
C7H15NaO3S
202.25
REFMEZARFCPESH-UHFFFAOYSA-M
sodium 1-heptanesulfonate,sodium heptane-1-sulfonate,1-heptanesulfonic acid sodium salt,1-heptanesulfonic acid, sodium salt,sodium heptane-1-sulphonate,1-heptanesulfonic acid, sodium salt 1:1,1-heptanesulfonic acid sodium,sodium heptanesulfonate,1-heptylsodiumsulfonate,sodium heptane sulfonate
23672332
sodium;heptane-1-sulfonate
CCCCCCCS(=O)(=O)[O-].[Na+]
Quantity 5g
Color White
Packaging Glass bottle
Loss on Drying 9% max. (100°C, 3 hrs) (vacuum)
Infrared Spectrum Authentic
Melting Point >300.0°C
Assay Percent Range 98%
Linear Formula CH3(CH2)6SO3Na
Beilstein 04, IV, 57
Solubility Information (10% in water) Clear colorless
Formula Weight 202.25
Physical Form Crystalline Powder of Flakes
Percent Purity 98%
Chemical Name or Material 1-Heptanesulfonic acid, sodium salt
Show More Show Less

GHS Signal Word: Warning

EINECSNumber : 245-210-5