Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Heptanesulfonic acid, sodium salt, 98%, ACROS Organics™
Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Glass Bottle
Specifications
Packaging | Glass Bottle |
Infrared Spectrum | Authentic |
Color | White |
Quantity | 25g |
Loss on Drying | 9% max. (100°C, 3 hrs) (vacuum) |
Assay Percent Range | 98% |
Linear Formula | CH3(CH2)6SO3Na |
Beilstein | 04, IV, 57 |
Solubility Information | Solubility in water: Soluble |
Formula Weight | 202.25 |
Show More |
Safety and Handling
Warning