Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Heptanesulfonic Acid Sodium Salt, HPLC Grade, J.T.Baker™
Click to view available options
Quantity:
25g
500g
Packaging:
Wide Mouth Amber Glass Bottle
Description
- For Ion-Pair Chromatography of Basic Compounds; For Laboratory,Research,or Manufacturing Use
Specifications
Packaging | Wide Mouth Amber Glass Bottle |
Quantity | 500g |
Specific Gravity | 1L = 0.00kg |
Formula Weight | 202.26 |
Grade | HPLC |
Chemical Name or Material | 1-Heptanesulfonic Acid Sodium Salt |