Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Heptanesulfonic Acid Sodium Salt, HPLC Grade, J.T.Baker™

Click to view available options
Quantity:
25g
500g
Packaging:
Wide Mouth Amber Glass Bottle
22767-50-6
C7H15NaO3S
202.244
REFMEZARFCPESH-UHFFFAOYSA-M
sodium 1-heptanesulfonate,sodium heptane-1-sulfonate,1-heptanesulfonic acid sodium salt,1-heptanesulfonic acid, sodium salt,sodium heptane-1-sulphonate,1-heptanesulfonic acid, sodium salt 1:1,1-heptanesulfonic acid sodium,sodium heptanesulfonate,1-heptylsodiumsulfonate,sodium heptane sulfonate
23672332
sodium;heptane-1-sulfonate
CCCCCCCS(=O)(=O)[O-].[Na+]
22767-50-6
C7H15NaO3S
202.244
REFMEZARFCPESH-UHFFFAOYSA-M
sodium 1-heptanesulfonate,sodium heptane-1-sulfonate,1-heptanesulfonic acid sodium salt,1-heptanesulfonic acid, sodium salt,sodium heptane-1-sulphonate,1-heptanesulfonic acid, sodium salt 1:1,1-heptanesulfonic acid sodium,sodium heptanesulfonate,1-heptylsodiumsulfonate,sodium heptane sulfonate
23672332
sodium;heptane-1-sulfonate
CCCCCCCS(=O)(=O)[O-].[Na+]

High purity solvents for use in analytical and preparative separations

    For Ion-Pair Chromatography of Basic Compounds; For Laboratory,Research,or Manufacturing Use
Packaging Wide Mouth Amber Glass Bottle
Quantity 25g
Specific Gravity 1L = 0.00kg
Formula Weight 202.26
Grade HPLC
Chemical Name or Material 1-Heptanesulfonic Acid Sodium Salt