Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Heptanesulfonic acid sodium salt monohydrate, HPLC grade, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatography
Specifications
Absorbance | (2.7g in 100mL water) (pH 3), (2.7 g in 100 ml water) (pH 3) at 0nm, 0.06 max. at 254nm, 0.06 max. at 280nm, 0.1 max. at 220nm, 1 max. at 210nm |
Melting Point | >300.0°C |
Color | White |
Loss on Drying | 7.4% to 9% (100°C, 3 hrs) (vacuum) |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Molecular Formula | C7H15NaO3S·H2O |
Linear Formula | CH3(CH2)6SO3Na·H2O |
Quantity | 5 g |
Solubility Information | (10% w/v in water) Clear colorless |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction