Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Heptanesulfonic Acid Sodium Salt, Spectrum™

Click to view available options
Quantity:
5 g
25 g
50 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
207300-90-1
207300-90-1
Packaging Amber Glass Bottle
Absorbance 0.05 (0.1 N Solution at 210 nm)
Quantity 1 kg
CAS Max % 1
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.