Alfa Aesar™ 1-Heptyl isocyanate, 98%

Manufacturer: Alfa Aesar™ B2179314

Quantity 25g

 View more versions of this product

Catalog No. AAB2179314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4747-81-3
    Molecular Formula CH3(CH2)6NCO
    Formula Weight 141.21
    MDL Number MFCD00144892
    UN Number UN2206
    Beilstein 1754817
    Chemical Name or Material 1-Heptyl isocyanate

    Specifications

    Formula Weight 141.21
    Odor Pungent
    UN Number UN2206
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H334-H227-H302-H315-H319-H335
    Toxic if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Combustible liquid.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Flash Point 62°C (143°F)
    Health Hazard 3 GHS P Statement
    P210-P284a-P261-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 25g
    Molecular Formula CH3(CH2)6NCO
    MDL Number MFCD00144892
    CAS 4747-81-3
    Beilstein 1754817
    Boiling Point 184°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
    Chemical Name or Material 1-Heptyl isocyanate
    Assay 98%

    GHS H Statement
    H331-H334-H227-H302-H315-H319-H335
    Toxic if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Combustible liquid.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P284a-P261-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit