Alfa Aesar™ (1-Heptyl)triphenylphosphonium bromide, 98+%

Manufacturer: Alfa Aesar™ B2542414

Quantity 25g

 View more versions of this product

Catalog No. AAB2542414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13423-48-8
    Molecular Formula C25H30BrP
    Linear Formula CH3(CH2)6P(C6H5)3Br
    Formula Weight 441.39
    MDL Number MFCD00050249
    Beilstein 3580843
    Chemical Name or Material (1-Heptyl)triphenylphosphonium bromide

    Specifications

    Formula Weight 441.39
    Odor Odorless
    Melting Point 176° to 180°C
    Quantity 25g
    Linear Formula CH3(CH2)6P(C6H5)3Br
    MDL Number MFCD00050249
    CAS 13423-48-8
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Molecular Formula C25H30BrP
    Beilstein 3580843
    Health Hazard 1 Warning
    Chemical Name or Material (1-Heptyl)triphenylphosphonium bromide
    Assay ≥98%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit