Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Heptyne, 99%

Manufacturer:  Alfa Aesar™ A1113006

Quantity 5g

 View more versions of this product

Catalog No. AAA1113006

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Applications
1-Heptyne is used in the synthesis of (6Z,10E,12Z)-octadeca-6,10,12-trienoic acid and (8Z,12E,14Z)-eicosa-8,12,14-trienoic acid.

Solubility
Soluble in organic solvents. Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Description & Specifications

Chemical Identifiers

628-71-7
96.17
UN3295
n-Pentylacetylene
CH3(CH2)4C≡CH
MFCD00009529
1733518
1-Heptyne

Specifications

5g
Unpleasant
96.17
UN3295
n-Pentylacetylene
−81°C
1-Heptyne
628-71-7
CH3(CH2)4C≡CH
MFCD00009529
1733518
98° to 100°C
−10°C (14°F)
99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.

GHS P Statement
P210-P243-P301+P330+P331-P315-P403+P233
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take precautionary measures against static discharge.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Get immediate medical advice/attention.
Store in a well-ventilated place.
Keep container tightly closed.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit