Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Heptyne, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
1-Heptyne is used in the synthesis of (6Z,10E,12Z)-octadeca-6,10,12-trienoic acid and (8Z,12E,14Z)-eicosa-8,12,14-trienoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Heptyne is used in the synthesis of (6Z,10E,12Z)-octadeca-6,10,12-trienoic acid and (8Z,12E,14Z)-eicosa-8,12,14-trienoic acid.
Solubility
Soluble in organic solvents. Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
1-Heptyne is used in the synthesis of (6Z,10E,12Z)-octadeca-6,10,12-trienoic acid and (8Z,12E,14Z)-eicosa-8,12,14-trienoic acid.
Solubility
Soluble in organic solvents. Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | -81°C |
Density | 0.736 |
Boiling Point | 98°C to 100°C |
Flash Point | −10°C (14°F) |
Odor | Unpleasant |
Refractive Index | 1.4095 |
Quantity | 25 g |
UN Number | UN3295 |
Beilstein | 1733518 |
Solubility Information | Soluble in organic solvents. Insoluble in water. |
Show More |
Safety and Handling
Hazard Category | H225-H304-H315-H319-H335 |
Hazard Statement | GHS H Statement H225-H304 Highly flammable liquid and vapor. May be fatal if swallowed and enters airways. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S. |
EINECSNumber | 211-051-5 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction