Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hexadecanol, 96%, ACROS Organics™

Click to view available options
Quantity:
1kg
2.5kg
10kg
36653-82-4
C16H34O
242.44
BXWNKGSJHAJOGX-UHFFFAOYSA-N
1-hexadecanol,cetyl alcohol,hexadecanol,cetanol,palmityl alcohol,hexadecyl alcohol,n-cetyl alcohol,cetaffine,cetylol,cetal
2682
CHEBI:16125
hexadecan-1-ol
CCCCCCCCCCCCCCCCO
36653-82-4
C16H34O
242.44
BXWNKGSJHAJOGX-UHFFFAOYSA-N
1-hexadecanol,cetyl alcohol,hexadecanol,cetanol,palmityl alcohol,hexadecyl alcohol,n-cetyl alcohol,cetaffine,cetylol,cetal
2682
CHEBI:16125
hexadecan-1-ol
CCCCCCCCCCCCCCCCO
Flash Point >157°C
Melting Point 47°C to 50°C
Boiling Point 344°C
Packaging Fibre drum
Viscosity 8 mPa.s (60°C)
Infrared Spectrum Authentic
Quantity 10kg
Color White
Assay Percent Range 96%
Linear Formula CH3(CH2)15OH
Beilstein 01,429
Merck Index 15,2027
Solubility Information Solubility in water: insoluble in water. Other solubilities: slightly soluble in ethanol,soluble in acetone,very soluble in ether,benzene,chloroform
Formula Weight 242.44
Physical Form Powder, Flakes or Pellets
Percent Purity ≥95%
Chemical Name or Material 1-Hexadecanol
Show More Show Less