Learn More
1-Hexadecanol, 98%, Thermo Scientific Chemicals
Description
1-Hexadecanol is used as cosmetic ingredient, emollient, foam stabilizer. It is also used as the raw material for the manufacture of surfactants. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Hexadecanol is used as cosmetic ingredient, emollient, foam stabilizer. It is also used as the raw material for the manufacture of surfactants. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
Solubility
Soluble in alcohol, chloroform, ether
Notes
Keep away from direct sunlight. Incompatible with strong oxidizing agents, strong acids. Store at 2-8°C.
Specifications
Melting Point | 47°C to 51°C |
Density | 0.817 g/mL |
Boiling Point | 190°C (15 mm) |
Flash Point | 135°C (275°F) |
Assay Percent Range | 98% |
Quantity | 500 g |
Beilstein | 1748475 |
Merck Index | 14,2028 |
Solubility Information | Soluble in alcohol, chloroform, ether |
Formula Weight | 242.45 |
Show More |
Safety and Handling
Hazard Category | H315-H319 |
Hazard Statement | GHS H Statement H315-H319-H335 |
Precautionary Statement | P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362 |
EINECSNumber | 253-149-0 |
RTECSNumber | MM0225000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.