Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hexanesulfonic Acid Sodium Salt, HPLC Grade, J.T.Baker™

Click to view available options
Quantity:
25g
500g
Packaging:
Wide Mouth Amber Glass Bottle
2832-45-3
C6H13NaO3S
188.217
QWSZRRAAFHGKCH-UHFFFAOYSA-M
sodium 1-hexanesulfonate,sodium hexane-1-sulfonate,sodium hexanesulfonate,1-hexanesulfonic acid sodium salt,1-hexanesulfonic acid, sodium salt,1-hexanesulfonic acid, sodium salt 1:1,1-hexane sulfonic acid sodium salt,hexyl sodium sulfonate,1-hexanesulfonate, sodium,hexanesulfonic acid na-salt
23677630
sodium;hexane-1-sulfonate
CCCCCCS(=O)(=O)[O-].[Na+]
2832-45-3
C6H13NaO3S
188.217
QWSZRRAAFHGKCH-UHFFFAOYSA-M
sodium 1-hexanesulfonate,sodium hexane-1-sulfonate,sodium hexanesulfonate,1-hexanesulfonic acid sodium salt,1-hexanesulfonic acid, sodium salt,1-hexanesulfonic acid, sodium salt 1:1,1-hexane sulfonic acid sodium salt,hexyl sodium sulfonate,1-hexanesulfonate, sodium,hexanesulfonic acid na-salt
23677630
sodium;hexane-1-sulfonate
CCCCCCS(=O)(=O)[O-].[Na+]

High purity solvents for use in analytical and preparative separations

    For Ion-Pair Chromatography of Basic Compounds
Specific Gravity 1L = 0.00kg
Packaging Wide Mouth Amber Glass Bottle
Quantity 25g
Formula Weight 188.22
Grade HPLC
Chemical Name or Material 1-Hexanesulfonic Acid Sodium Salt