Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-hexanesulfonic acid, sodium salt hydrate, 99%, ACROS Organics™
Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
Specifications
Density | Negligible |
Color | White |
Absorbance | 5.5 w/v% aq. soln., pH 3, H3PO4,0.06 max. at 280nm,0.1 max. at 220nm,1 max. at 210nm,5.5 w/v% aq. soln., pH 3, H3PO4 at 0nm |
Quantity | 5g |
Melting Point | >300.0°C |
Infrared Spectrum | Authentic |
Loss on Drying | 9.6% max. (100°C, 3 hrs) (vacuum) |
Vapor Pressure | Negligible |
Packaging | Glass bottle |
Assay Percent Range | 99% |
Show More |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath
GHS Signal Word: Warning