Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hexanesulfonic Acid Sodium Salt, Monohydrate, 98%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
207300-91-2
207300-91-2
Packaging Amber Glass Bottle
Quantity 1 kg
CAS Max % 1
Percent Purity 0.98
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.