Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Hexanol, ≥99.9% (GC), MilliporeSigma™ Supelco™
$50.25 - $208.00
Specifications
CAS | 111-27-3 |
---|---|
Type | Analytical Standard |
Linear Formula | CH3(CH2)5 OH |
MDL Number | MFCD00002982 |
Synonym | Hexyl alcohol |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
11-102-0401
|
MilliporeSigma Supelco
731171MLF |
1 mL |
Each for $50.25
|
|
|||||
11-101-8781
|
MilliporeSigma Supelco
731175MLF |
5 mL |
Each for $208.00
|
|
|||||
Description
Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal CareSpecifications
111-27-3 | |
CH3(CH2)5 OH | |
Hexyl alcohol | |
≥99.9% (GC) | |
559°F | |
4.5 (vs. air) | |
-52°C | |
60°C |
Analytical Standard | |
MFCD00002982 | |
102.17 | |
Analytical Standard | |
1 mmHg (25.6°C) | |
0.814 g/mL (at 25°C) | |
156°C to 157°C | |
Neat |
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label
RTECSNumber : MQ4025000
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title