Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hexene, 97%, ACROS Organics™

Click to view available options
Quantity:
100mL
1L
2.5L
25L
Packaging:
Glass bottle
Metal Drum
592-41-6
C6H12
84.15
MFCD00009505
LIKMAJRDDDTEIG-UHFFFAOYSA-N
1-hexene,hexene,butylethylene,hexylene,hexene-1,1-n-hexene,butyl ethylene,dialene 6,n-hexene,unii-b38zz8c206
11597
CHEBI:24579
hex-1-ene
CCCCC=C
592-41-6
C6H12
84.15
MFCD00009505
LIKMAJRDDDTEIG-UHFFFAOYSA-N
1-hexene,hexene,butylethylene,hexylene,hexene-1,1-n-hexene,butyl ethylene,dialene 6,n-hexene,unii-b38zz8c206
11597
CHEBI:24579
hex-1-ene
CCCCC=C

1-Hexene, 97%, C6H12, CAS Number-592-41-6, dialene 6, butyl ethylene, 1-hexene, butylethylene, 1-n-hexene, hexylene, hexene-1, hexene, n-hexene, unii-b38zz8c206, 1L, 96% min. (GC), 01, 215, 62.0 deg.C to 65.0 deg.C, 100.0, 96.0, CHEBI:24579, Colorless, 0.6780g/mL, 209-753-1, -26 deg.C, 84.15

1-Hexene, 97%, C6H12, CAS Number-592-41-6, dialene 6, butyl ethylene, 1-hexene, butylethylene, 1-n-hexene, hexylene, hexene-1, hexene, n-hexene, unii-b38zz8c206, 1L, 96% min. (GC), 01, 215, 62.0 deg.C to 65.0 deg.C, 100.0, 96.0, CHEBI:24579, Colorless, 0.6780g/mL, 209-753-1, -26 deg.C, 84.15

Specific Gravity 0.678
Refractive Index 1.3867 to 1.3887
Melting Point -140.0°C
Viscosity 0.34 mm2/s (40°C)
Boiling Point 62.0°C to 65.0°C
Density 0.6780g/mL
Color Colorless
Quantity 25L
Infrared Spectrum Authentic
Flash Point −26°C
Packaging Metal Drum
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula CH3(CH2)3CH=CH2
Beilstein 01, 215
Solubility Information Solubility in water: 50 mg/l (20°C). Other solubilities: soluble in alcohol and hydrocarbon solvents, soluble in ethanol and diethyl ether
Formula Weight 84.15
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 1-Hexene
Show More Show Less

GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take precautionary measures against static discharge.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Imm

GHS Signal Word: Danger

EINECSNumber : 209-753-1