Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hexyl-2,3-dimethylimidazolium chloride, 97%, Acros Organics

Click to view available options
Quantity:
25g
455270-59-4
C11H21ClN2
216.753
MFCD03093292
NCTQEXWJEPRHLV-UHFFFAOYSA-M
1-hexyl-2,3-dimethylimidazolium chloride,1-hexyl-2,3-dimethyl-1h-imidazol-3-ium chloride,ksc496i5f,1-hexyl-2,3-dimethylimidazol-3-ium chloride,1-hexyl-2,3-dimethylimidazol-1-ium chloride
2734169
1-hexyl-2,3-dimethylimidazol-3-ium;chloride
CCCCCCN1C=C[N+](=C1C)C.[Cl-]
455270-59-4
C11H21ClN2
216.753
MFCD03093292
NCTQEXWJEPRHLV-UHFFFAOYSA-M
1-hexyl-2,3-dimethylimidazolium chloride,1-hexyl-2,3-dimethyl-1h-imidazol-3-ium chloride,ksc496i5f,1-hexyl-2,3-dimethylimidazol-3-ium chloride,1-hexyl-2,3-dimethylimidazol-1-ium chloride
2734169
1-hexyl-2,3-dimethylimidazol-3-ium;chloride
CCCCCCN1C=C[N+](=C1C)C.[Cl-]
Ionic liquid
Quantity 25g
Color Yellow
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 216.75
Physical Form Lumpy Solid
Percent Purity ≥96.0%
Chemical Name or Material 1-Hexyl-2, 3-dimethylimidazolium chloride

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC