Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hexylpyridinium chloride, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
6220-15-1
C11H18ClN
199.72
MFCD03093294
JEOSMYVMLZTQOH-UHFFFAOYSA-M
1-hexylpyridinium chloride,1-hexylpyridin-1-ium chloride,hexylpyridinium chloride,1-hexylpyridiniumchloride,dsstox_cid_27911,dsstox_rid_82664,dsstox_gsid_47935
2734172
1-hexylpyridin-1-ium;chloride
CCCCCC[N+]1=CC=CC=C1.[Cl-]
6220-15-1
C11H18ClN
199.72
MFCD03093294
JEOSMYVMLZTQOH-UHFFFAOYSA-M
1-hexylpyridinium chloride,1-hexylpyridin-1-ium chloride,hexylpyridinium chloride,1-hexylpyridiniumchloride,dsstox_cid_27911,dsstox_rid_82664,dsstox_gsid_47935
2734172
1-hexylpyridin-1-ium;chloride
CCCCCC[N+]1=CC=CC=C1.[Cl-]
Ionic liquid
Quantity 25g
Infrared Spectrum Authentic
Color Brown
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (Argentometry)
Formula Weight 199.72
Physical Form Fused Solid
Percent Purity 97%
Chemical Name or Material 1-Hexylpyridinium chloride
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning