Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hexylpyridinium hexafluorophosphate, 97+%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
797789-00-5
C11H18F6NP
309.236
MFCD03093298
DKRTWJPQQYAIHH-UHFFFAOYSA-N
1-hexylpyridinium hexafluorophosphate,1-hexylpyridin-1-ium hexafluorophosphate v,1-hexylpyridin-1-ium hexafluorophosphate,dsstox_cid_27912,dsstox_rid_82665,dsstox_gsid_47936,ksc376g7d,n-hexylpyridinium hexafluorophosphate
2734177
1-hexylpyridin-1-ium;hexafluorophosphate
CCCCCC[N+]1=CC=CC=C1.F[P-](F)(F)(F)(F)F
797789-00-5
C11H18F6NP
309.236
MFCD03093298
DKRTWJPQQYAIHH-UHFFFAOYSA-N
1-hexylpyridinium hexafluorophosphate,1-hexylpyridin-1-ium hexafluorophosphate v,1-hexylpyridin-1-ium hexafluorophosphate,dsstox_cid_27912,dsstox_rid_82665,dsstox_gsid_47936,ksc376g7d,n-hexylpyridinium hexafluorophosphate
2734177
1-hexylpyridin-1-ium;hexafluorophosphate
CCCCCC[N+]1=CC=CC=C1.F[P-](F)(F)(F)(F)F
Ionic liquid
Melting Point 45°C to 48°C
NMR 97.0% min.
Quantity 25g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Beige
CAS Min % 97.0
CAS Max % 100.0
Assay Percent Range 97+%
Formula Weight 309.23
Physical Form Crystals
Chemical Name or Material 1-Hexylpyridinium hexafluorophosphate
Show More Show Less

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger