Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Hydrazinophthalazine hydrochloride, 98%

Manufacturer:  Alfa Aesar™ B2299514

Quantity 25g

 View more versions of this product

Catalog No. AAB2299514


  Check Availability
Add to cart

Specifications

Description

Applications
A monoamine oxidase inhibitor1-Hydrazinophthalazine hydrochloride is a non-nucleoside analog that inhibits DNA methylation and reactivates the expression of tumor suppressor genes. It acts as a non-selective MAO-A/B inhibitor and semicarbazide sensitive amine oxidase inhibitor. Further, it serves as an antihypertensive.

Solubility
Soluble in water. Slightly soluble in diethyl ether and alcohol.

Notes
Incompatible with strong oxidizing agents.
Description & Specifications

Chemical Identifiers

304-20-1
196.64
UN2811
14,4763
1-Hydrazinophthalazine hydrochloride
C8H8N4·HCl
MFCD00135998
3568329
Hydralazine hydrochloride; 1(2H)-Phthalazinone hydrazone

Specifications

≈275°C (dec.)
304-20-1
196.64
UN2811
14,4763
1-Hydrazinophthalazine hydrochloride
25g
C8H8N4·HCl
MFCD00135998
3568329
Hydralazine hydrochloride; 1(2H)-Phthalazinone hydrazone
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H351-H361
Toxic if swallowed.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.

GHS P Statement
P281-P301+P310-P308+P313-P321-P405-P501a
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF exposed or concerned: Get medical advice/attention.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : TH9000000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit