Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Hydrazinophthalazine hydrochloride, 98%

Click to view available options
Quantity:
5g
304-20-1
C8H9ClN4
196.638
MFCD00135998
ZUXNZUWOTSUBMN-UHFFFAOYSA-N
hydralazine hydrochloride,1-hydrazinophthalazine hydrochloride,1-hydrazinylphthalazine hydrochloride,apresoline,aiselazine,hydralazine hcl,hydralazine chloride,hyperazin,hyperex,lopress
9351
CHEBI:31672
phthalazin-1-ylhydrazine;hydrochloride
C1=CC=C2C(=C1)C=NN=C2NN.Cl
304-20-1
C8H9ClN4
196.638
MFCD00135998
ZUXNZUWOTSUBMN-UHFFFAOYSA-N
hydralazine hydrochloride,1-hydrazinophthalazine hydrochloride,1-hydrazinylphthalazine hydrochloride,apresoline,aiselazine,hydralazine hcl,hydralazine chloride,hyperazin,hyperex,lopress
9351
CHEBI:31672
phthalazin-1-ylhydrazine;hydrochloride
C1=CC=C2C(=C1)C=NN=C2NN.Cl
Applications
A monoamine oxidase inhibitor1-Hydrazinophthalazine hydrochloride is a non-nucleoside analog that inhibits DNA methylation and reactivates the expression of tumor suppressor genes. It acts as a non-selective MAO-A/B inhibitor and semicarbazide sensitive amine oxidase inhibitor. Further, it serves as an antihypertensive.

Solubility
Soluble in water. Slightly soluble in diethyl ether and alcohol.

Notes
Incompatible with strong oxidizing agents.
Quantity 5g
Melting Point ~275°C (decomposition)
UN Number UN2811
Beilstein 3568329
Merck Index 14,4763
Formula Weight 196.64
Percent Purity 98%
Chemical Name or Material 1-Hydrazinophthalazine hydrochloride

GHS H Statement
H301-H351-H361
Toxic if swallowed.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.

GHS P Statement
P281-P301+P310-P308+P313-P321-P405-P501a
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF exposed or concerned: Get medical advice/attention.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : TH9000000

TSCA : TSCA