Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hydroxy-2,2,4,5,5-pentamethyl-3-imidazoline-3-oxide, ACROS Organics™

Click to view available options
Quantity:
100mg
18796-02-6
C8H16N2O2
172.228
MFCD00142634
ANKGMEQLPXFSPL-UHFFFAOYSA-N
2,2,4,5,5-pentamethyl-1-imidazolidinol 3-oxide,2,2,4,5,5-pentamethyl-2,5-dihydro-1h-imidazol-1-ol 3-oxide,2,2,4,5,5-pentamethyl-3-oxy-2,5-dihydro-imidazol-1-ol,3-hydroxy-2,2,4,4,5-pentamethylimidazol-1-ium-1-olate,1-hydroxy-2,2,4,5,5-pentamethyl-3-imidazoline 3-oxide,1-hydroxy-2,2,4,5,5-pentamethyl-3-imidazoline-3-oxide,3-hydroxy-2,2,4,4,5-pentamethyl-1-oxido-imidazol-1-ium,2,2,4,4,5-pentamethyl-1-oxidanidyl-3-oxidanyl-imidazol-1-ium,3-hydroxy-2,2,4,4,5-pentamethyl-1-oxido-3-imidazolin-1-ium,2,2,4,5,5-pentamethyl-3-oxidoimidazol-3-ium-1-yloxidanyl
379371
3-hydroxy-2,2,4,4,5-pentamethyl-1-oxidoimidazol-1-ium
CC1=[N+](C(N(C1(C)C)O)(C)C)[O-]
18796-02-6
C8H16N2O2
172.228
MFCD00142634
ANKGMEQLPXFSPL-UHFFFAOYSA-N
2,2,4,5,5-pentamethyl-1-imidazolidinol 3-oxide,2,2,4,5,5-pentamethyl-2,5-dihydro-1h-imidazol-1-ol 3-oxide,2,2,4,5,5-pentamethyl-3-oxy-2,5-dihydro-imidazol-1-ol,3-hydroxy-2,2,4,4,5-pentamethylimidazol-1-ium-1-olate,1-hydroxy-2,2,4,5,5-pentamethyl-3-imidazoline 3-oxide,1-hydroxy-2,2,4,5,5-pentamethyl-3-imidazoline-3-oxide,3-hydroxy-2,2,4,4,5-pentamethyl-1-oxido-imidazol-1-ium,2,2,4,4,5-pentamethyl-1-oxidanidyl-3-oxidanyl-imidazol-1-ium,3-hydroxy-2,2,4,4,5-pentamethyl-1-oxido-3-imidazolin-1-ium,2,2,4,5,5-pentamethyl-3-oxidoimidazol-3-ium-1-yloxidanyl
379371
3-hydroxy-2,2,4,4,5-pentamethyl-1-oxidoimidazol-1-ium
CC1=[N+](C(N(C1(C)C)O)(C)C)[O-]
Color White
Melting Point 180°C to 188°C
Quantity 100mg
Infrared Spectrum Authentic
Formula Weight 172.23
Physical Form Crystals and Needles
Chemical Name or Material 1-Hydroxy-2,2,4,5,5-pentamethyl-3-imidazoline-3-oxide