Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hydroxy-2-naphthoic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
Packaging:
Glass bottle
Plastic bottle
86-48-6
C11H8O3
188.18
MFCD00003960
SJJCQDRGABAVBB-UHFFFAOYSA-N
1-hydroxy-2-naphthoic acid,1-naphthol-2-carboxylic acid,2-carboxy-1-naphthol,2-naphthalenecarboxylic acid, 1-hydroxy,1-hydroxy-2-naphthoate,1-hydroxy-2-naphthalenecarboxylic acid,unii-u8lz3r07l8,2-naphthoic acid, 1-hydroxy,alpha-hydroxynaphthoic acid,1-hydroxy-naphthalene-2-carboxylic acid
6844
CHEBI:36108
1-hydroxynaphthalene-2-carboxylic acid
C1=CC=C2C(=C1)C=CC(=C2O)C(=O)O
86-48-6
C11H8O3
188.18
MFCD00003960
SJJCQDRGABAVBB-UHFFFAOYSA-N
1-hydroxy-2-naphthoic acid,1-naphthol-2-carboxylic acid,2-carboxy-1-naphthol,2-naphthalenecarboxylic acid, 1-hydroxy,1-hydroxy-2-naphthoate,1-hydroxy-2-naphthalenecarboxylic acid,unii-u8lz3r07l8,2-naphthoic acid, 1-hydroxy,alpha-hydroxynaphthoic acid,1-hydroxy-naphthalene-2-carboxylic acid
6844
CHEBI:36108
1-hydroxynaphthalene-2-carboxylic acid
C1=CC=C2C(=C1)C=CC(=C2O)C(=O)O
Infrared Spectrum Authentic
Flash Point >150°C
Packaging Glass bottle
Quantity 5g
Melting Point 199.0°C to 205.0°C
Color Tan to White
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 10, 331
Merck Index 15, 4873
Solubility Information Solubility in water: soluble in hot water. Other solubilities: practically insoluble in cold water, soluble in benzene and chloroform, freely soluble in alcohol and ether
Formula Weight 188.18
Physical Form Powder or Crystals
Percent Purity 98%
Chemical Name or Material 1-Hydroxy-2-naphthoic acid, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 201-674-0

TSCA : TSCA