Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hydroxy-7-azabenzotriazole 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
39968-33-7
C5H4N4O
136.114
FPIRBHDGWMWJEP-UHFFFAOYSA-N
1-hydroxy-7-azabenzotriazole,hoat,3h-1,2,3 triazolo 4,5-b pyridin-3-ol,3-hydroxytriazolo 4,5-b pyridine,unii-tx8xyh09h0,1-hydroxy-7-azabenzotriazole hoat,hoat 1-hydroxy-7-azabenzotriazole,3h-1,2,3-triazolo 4,5-b pyridine, 3-hydroxy,tx8xyh09h0,7-aza-1-hydroxybenzotriazole
181649
3-hydroxytriazolo[4,5-b]pyridine
C1=CC2=C(N=C1)N(N=N2)O
39968-33-7
C5H4N4O
136.114
FPIRBHDGWMWJEP-UHFFFAOYSA-N
1-hydroxy-7-azabenzotriazole,hoat,3h-1,2,3 triazolo 4,5-b pyridin-3-ol,3-hydroxytriazolo 4,5-b pyridine,unii-tx8xyh09h0,1-hydroxy-7-azabenzotriazole hoat,hoat 1-hydroxy-7-azabenzotriazole,3h-1,2,3-triazolo 4,5-b pyridine, 3-hydroxy,tx8xyh09h0,7-aza-1-hydroxybenzotriazole
181649
3-hydroxytriazolo[4,5-b]pyridine
C1=CC2=C(N=C1)N(N=N2)O
Quantity 5g
CAS Min % 97.5
CAS Max % 100.0
Formula Weight 136.11
Chemical Name or Material 1-Hydroxy-7-azabenzotriazole, 98%