Alfa Aesar™ 1-Hydroxyethylidenebis(phosphonic acid), 96%

Manufacturer: Alfa Aesar™ 4350204

Quantity 2g

 View more versions of this product

Catalog No. AA4350204

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2809-21-4
    Molecular Formula CH3C(OH)(PO3H2)2
    Formula Weight 206.03
    MDL Number MFCD00070585
    UN Number UN3261
    Merck Index 14,3863
    Synonym Etidronic acid; 1-Hydroxyethane-1,1-diphosphonic acid
    Chemical Name or Material 1-Hydroxyethylidenebis (phosphonic acid)

    Specifications

    Formula Weight 206.03
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Molecular Formula CH3C(OH)(PO3H2)2
    MDL Number MFCD00070585
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    CAS 2809-21-4
    UN Number UN3261
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Physical Form Powder
    Merck Index 14,3863
    Synonym Etidronic acid; 1-Hydroxyethane-1,1-diphosphonic acid
    RTECS Number SZ8562100
    Odor Odorless
    Quantity 2g
    Chemical Name or Material 1-Hydroxyethylidenebis (phosphonic acid)
    Assay 96%

    GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit