Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(γ-Hydroxypropyl)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride, Acros Organics

Click to view available options
Quantity:
500mg
51452-52-9
C14H22ClNO3
287.784
MFCD01863271
VXJZIWCLBQUDKP-UHFFFAOYSA-N
1-gamma-hydroxypropyl-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride,3-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-1-yl propan-1-ol hydrochloride
90473071
3-(6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-1-yl)propan-1-ol;hydrochloride
COC1=C(C=C2C(NCCC2=C1)CCCO)OC.Cl
51452-52-9
C14H22ClNO3
287.784
MFCD01863271
VXJZIWCLBQUDKP-UHFFFAOYSA-N
1-gamma-hydroxypropyl-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride,3-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-1-yl propan-1-ol hydrochloride
90473071
3-(6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-1-yl)propan-1-ol;hydrochloride
COC1=C(C=C2C(NCCC2=C1)CCCO)OC.Cl
Quantity 500mg
Formula Weight 287.79
Chemical Name or Material 1-(γ-Hydroxypropyl)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride