Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Hydroxytetraphenylcyclopentadienyl(tetraphenyl-2,4-cyclopentadien-1-one)-mu-hydrotetracarbonyldiruthenium(II), 98%

Manufacturer:  Alfa Aesar™ 4688402

Quantity 0.5g

 View more versions of this product

Catalog No. AA4688402


  Check Availability
Add to cart

Specifications

Description

  • Calayst for hydrogen transfers including reduction of ketones and aldehydes, and oxidation and/or racemization of amines and alcohols
Description & Specifications

Chemical Identifiers

104439-77-2
1085.15
14,8483
1-Hydroxytetraphenylcyclopentadienyl (tetraphenyl-2,4-cyclopentadien-1-one)-mu-hydrotetracarbonyldiruthenium (II)
C62H42O6Ru2
MFCD03840556
Shvo's Catalyst

Specifications

Moisture Sensitive
223° to 227°C
104439-77-2
C62H42O6Ru2
MFCD03840556
Shvo's Catalyst
98%
Yellow to orange
0.5g
Crystalline
1085.15
14,8483
1-Hydroxytetraphenylcyclopentadienyl (tetraphenyl-2,4-cyclopentadien-1-one)-mu-hydrotetracarbonyldiruthenium (II)
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit