Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Indanol, 98%

Click to view available options
Quantity:
5g
25g
50g
6351-10-6
C9H10O
134.18
MFCD00003797
YIAPLDFPUUJILH-UHFFFAOYNA-N
1-indanol,indanol,indan-1-ol,1-hydroxyhydrindene,1h-inden-1-ol, 2,3-dihydro,1h-indenol, 2,3-dihydro,1-hydroxyindan,1-hydroxyindane,1-indanole,--indanol
22819
CHEBI:16697
2,3-dihydro-1H-inden-1-ol
C1CC2=CC=CC=C2C1O
6351-10-6
C9H10O
134.18
MFCD00003797
YIAPLDFPUUJILH-UHFFFAOYNA-N
1-indanol,indanol,indan-1-ol,1-hydroxyhydrindene,1h-inden-1-ol, 2,3-dihydro,1h-indenol, 2,3-dihydro,1-hydroxyindan,1-hydroxyindane,1-indanole,--indanol
22819
CHEBI:16697
2,3-dihydro-1H-inden-1-ol
C1CC2=CC=CC=C2C1O
Applications
1-Indanol derivative was found to be an efficient ligand for the palladium-catalyzed asymmetric Heck reaction. It is used as pharmaceutical intermediate.

Solubility
Soluble in water 10 g/L at 20°C, ethanol, benzene.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Quantity 25g
Flash Point 145°C (293°F)
Boiling Point 128°C (12mmHg)
Melting Point 50°C to 54°C
Beilstein 2042960
Formula Weight 134.18
Percent Purity 98%
Chemical Name or Material 1-Indanol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning