Alfa Aesar™ 1-Indanol, 98%

Manufacturer: Alfa Aesar™ B2453618

Quantity 50g

 View more versions of this product

Catalog No. AAB2453618

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6351-10-6
    Molecular Formula C9H10O
    Formula Weight 134.18
    MDL Number MFCD00003797
    Beilstein 2042960
    Synonym 1-Hydroxyindane
    Chemical Name or Material 1-Indanol

    Specifications

    Formula Weight 134.18
    Molecular Formula C9H10O
    Beilstein 2042960
    Synonym 1-Hydroxyindane
    Flash Point 145°C (293°F)
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00003797
    CAS 6351-10-6
    Health Hazard 1 Warning
    Boiling Point 128°C (12mmHg)
    Melting Point 50° to 54°C
    Quantity 50g
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 1-Indanol
    Assay 98%

    Description

    Applications
    1-Indanol derivative was found to be an efficient ligand for the palladium-catalyzed asymmetric Heck reaction. It is used as pharmaceutical intermediate.

    Solubility
    Soluble in water 10 g/L at 20°C, ethanol, benzene.

    Notes
    Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit