Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Indanol, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
6351-10-6
C9H10O
134.18
MFCD00003797
YIAPLDFPUUJILH-UHFFFAOYNA-N
1-indanol,indanol,indan-1-ol,1-hydroxyhydrindene,1h-inden-1-ol, 2,3-dihydro,1h-indenol, 2,3-dihydro,1-hydroxyindan,1-hydroxyindane,1-indanole,--indanol
22819
CHEBI:16697
2,3-dihydro-1H-inden-1-ol
C1CC2=CC=CC=C2C1O
6351-10-6
C9H10O
134.18
MFCD00003797
YIAPLDFPUUJILH-UHFFFAOYNA-N
1-indanol,indanol,indan-1-ol,1-hydroxyhydrindene,1h-inden-1-ol, 2,3-dihydro,1h-indenol, 2,3-dihydro,1-hydroxyindan,1-hydroxyindane,1-indanole,--indanol
22819
CHEBI:16697
2,3-dihydro-1H-inden-1-ol
C1CC2=CC=CC=C2C1O
Packaging Glass bottle
Boiling Point 260°C
Flash Point 145°C
Color White to Yellow
Quantity 10g
Melting Point 50°C to 54°C
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Beilstein 06, 574
Solubility Information Solubility in water: 10g/L (20°C). Other solubilities: soluble in ethanol,benzene and chloroform,slightly soluble in petroleum ether
Formula Weight 134.18
Physical Form Crystalline Solid
Percent Purity 98%
Chemical Name or Material 1-Indanol, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 228-755-3

TSCA : TSCA