Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Indanone, 99+%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
500g
83-33-0
C9H8O
132.16
QNXSIUBBGPHDDE-UHFFFAOYSA-N
1-indanone,indan-1-one,2,3-dihydro-1h-inden-1-one,indanone,alpha-indanone,alpha-hydrindone,1h-inden-1-one, 2,3-dihydro,1-indone,hydrindone,.alpha.-indanone
6735
CHEBI:17404
2,3-dihydroinden-1-one
C1CC(=O)C2=CC=CC=C21
83-33-0
C9H8O
132.16
QNXSIUBBGPHDDE-UHFFFAOYSA-N
1-indanone,indan-1-one,2,3-dihydro-1h-inden-1-one,indanone,alpha-indanone,alpha-hydrindone,1h-inden-1-one, 2,3-dihydro,1-indone,hydrindone,.alpha.-indanone
6735
CHEBI:17404
2,3-dihydroinden-1-one
C1CC(=O)C2=CC=CC=C21
Melting Point 38.0°C to 42.0°C
Quantity 500g
Packaging Glass Bottle
Density 1.1000g/mL
Flash Point >110°C
Boiling Point 243.0°C to 245.0°C
Specific Gravity 1.1
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Beilstein 07,36
Solubility Information Solubility in water: 6.5g/L (20°C). Other solubilities: soluble in ethanol,ether and chloroform
Formula Weight 132.16
Physical Form Crystalline Mass
Percent Purity 99+%
Chemical Name or Material 1-Indanone
Show More Show Less

GHS H Statement
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wash face,hands and any exposed skin thoroughly after handling.
Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 201-470-1