Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Iodo-2,4-dinitrobenzene, 98%

Click to view available options
Quantity:
5g
25g
709-49-9
C6H3IN2O4
294.004
MFCD00039738
FXMKXMJLXRTQSW-UHFFFAOYSA-N
2,4-dinitroiodobenzene,benzene, 1-iodo-2,4-dinitro,unii-not697x2xm,ccris 7907,4-iodo-1,3-dintrobenzene,6-iodo-1,3-dintrobenzene,2,4-dinitro-1-iodobenzene,not697x2xm,1-iodo-2,4-dinitro-benzene,1-iod-2,4-dinitrobenzol
69730
CHEBI:59000
1-iodo-2,4-dinitrobenzene
C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])I
709-49-9
C6H3IN2O4
294.004
MFCD00039738
FXMKXMJLXRTQSW-UHFFFAOYSA-N
2,4-dinitroiodobenzene,benzene, 1-iodo-2,4-dinitro,unii-not697x2xm,ccris 7907,4-iodo-1,3-dintrobenzene,6-iodo-1,3-dintrobenzene,2,4-dinitro-1-iodobenzene,not697x2xm,1-iodo-2,4-dinitro-benzene,1-iod-2,4-dinitrobenzol
69730
CHEBI:59000
1-iodo-2,4-dinitrobenzene
C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])I
Applications
It is employed in the cloning and characterization of a biotic-stress-inducible glutathione transferase from Phaseolus vulgaris.

Solubility
Insoluble in water.

Notes
Light sensitive. Store away from oxidizing agents, bases and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under dark.
Melting Point 87°C to 90°C
Quantity 5g
Beilstein 1113584
Formula Weight 294.01
Percent Purity 98%
Chemical Name or Material 1-Iodo-2,4-dinitrobenzene

GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P280g-P262-P305+P351+P338
Wear protective gloves.
Do not get in eyes, on skin, or on clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning