Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Iodo-2-(trimethylsilyl)acetylene, 97%

Manufacturer:  Alfa Aesar™ H5337503

Quantity 1g

 View more versions of this product

Catalog No. AAH5337503


  Check Availability
Add to cart

Specifications

Description

Applications
1-Iodo-2-(trimethylsilyl)acetylene may be used to investigate the reactivity of diamagnetic cobalt(I) complex, Tol-BDI((2-pp)2)Co [Tol-BDI((2-pp)2)H = 2-(4-tolyl)-1,3-bis(2-isopropylpyridyl)propenediimine]. It may be used for the synthesis of 1-(4-trimethylsilanylbuta-1,3-diynyl)pyrene, via Pd-coupling with 1-iodo-2-trimethylsilylacetylene.

Notes
Light sensitive. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

18163-47-8
(CH3)3SiC≡CI
MFCD00274201
1-Iodo-2-(trimethylsilyl)acetylene
C5H9ISi
224.12
UN1993

Specifications

18163-47-8
(CH3)3SiC≡CI
MFCD00274201
130°C
48°C (118°F)
97%
C5H9ISi
224.12
UN1993
1g
1-Iodo-2-(trimethylsilyl)acetylene

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H311-H334-H361-H226-H332-H317
Toxic if swallowed.
Toxic in contact with skin.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility or the unborn child.
Flammable liquid and vapor.
Harmful if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P210-P301+P310-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit