Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Iodo-3,5-bis(trifluoromethyl)benzene, 97+%

Manufacturer:  Alfa Aesar™ A1087822

Quantity 100g

 View more versions of this product

Catalog No. AAA1087822


  Check Availability
Add to cart

Specifications

Description

Applications
3,5-bis(trifluoromethyl)pheny1- magnesium iodide was prepared from 1-iodo-3,5- bis(trifluoromethyl)benzene. Monodeprotected masked hexayne was not particularly stable and hence was used directly without purification in a Sonogashira coupling with 1-iodo-3,5-bis(trifluoromethyl)benzene to provide mono-aryl-end-capped masked hexayne.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
Description & Specifications

Chemical Identifiers

328-73-4
340.01
2461627
1-Iodo-3,5-bis (trifluoromethyl)benzene
C8H3F6I
MFCD00040837
3,5-Bis(trifluoromethyl)iodobenzene

Specifications

328-73-4
340.01
2461627
3,5-Bis(trifluoromethyl)iodobenzene
74°C (165°F)
≥97%
C8H3F6I
MFCD00040837
62° to 64°C (12mmHg)
100g
1-Iodo-3,5-bis (trifluoromethyl)benzene

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit