Alfa Aesar™ 1-Iodo-3,5-dinitrobenzene, 99%

Manufacturer: Alfa Aesar™ H2817006

Quantity 5g

 View more versions of this product

Catalog No. AAH2817006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6276-04-6
    Molecular Formula C6H3IN2O4
    Formula Weight 294.01
    MDL Number MFCD00024330
    Synonym 3,5-Dinitroiodobenzene
    Chemical Name or Material 1-Iodo-3,5-dinitrobenzene

    Specifications

    Formula Weight 294.01
    Molecular Formula C6H3IN2O4
    MDL Number MFCD00024330
    Melting Point 99° to 103°C
    Synonym 3,5-Dinitroiodobenzene
    Quantity 5g
    Health Hazard 2 GHS H Statement
    H302-H312-H332-H315-H319-H317-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.
    CAS 6276-04-6
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P280h-P262-P305+P351+P338
    Wear protective gloves/clothing.
    Do not get in eyes, on skin, or on clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 1-Iodo-3,5-dinitrobenzene
    Assay 99%

    Description

    Applications
    As cytochemical reagent. For kinetic-potentiometric study and analytical applications of micellar-catalysed reactions.

    Solubility
    Slightly soluble in water.

    Notes
    Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.

    GHS H Statement
    H302-H312-H332-H315-H319-H317-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P262-P305+P351+P338
    Wear protective gloves/clothing.
    Do not get in eyes, on skin, or on clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit