Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Iodo-3-nitrobenzene, 99%

Click to view available options
Quantity:
25g
100g
645-00-1
C6H4INO2
249.007
MFCD00007218
CBYAZOKPJYBCHE-UHFFFAOYSA-N
3-iodonitrobenzene,m-iodonitrobenzene,benzene, 1-iodo-3-nitro,3-nitroiodobenzene,meta-iodonitrobenzene,3-i-nitrobenzene,iodo-3-nitrobenzene,meta-nitroiodobenzene,1-iodo-3-nitro-benzene,3-nitro-iodobenzen
12574
CHEBI:67123
1-iodo-3-nitrobenzene
C1=CC(=CC(=C1)I)[N+](=O)[O-]
645-00-1
C6H4INO2
249.007
MFCD00007218
CBYAZOKPJYBCHE-UHFFFAOYSA-N
3-iodonitrobenzene,m-iodonitrobenzene,benzene, 1-iodo-3-nitro,3-nitroiodobenzene,meta-iodonitrobenzene,3-i-nitrobenzene,iodo-3-nitrobenzene,meta-nitroiodobenzene,1-iodo-3-nitro-benzene,3-nitro-iodobenzen
12574
CHEBI:67123
1-iodo-3-nitrobenzene
C1=CC(=CC(=C1)I)[N+](=O)[O-]
Flash Point 71°C (159°F)
Melting Point 35°C to 38°C
Boiling Point 279°C to 280°C
Quantity 100g
UN Number UN2926
Beilstein 1525167
Formula Weight 249.01
Percent Purity 99%
Chemical Name or Material 1-Iodo-3-nitrobenzene

GHS H Statement
H331-H228-H373-H302-H312-H315-H319-H335
Toxic if inhaled.
Flammable solid.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P260-P261-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, TOXIC, ORGANIC, N.O.S.

TSCA : TSCA