Alfa Aesar™ 1-Iodo-3-(trifluoromethoxy)benzene, 97+%

Manufacturer: Alfa Aesar™ B2416903

Quantity 1g

 View more versions of this product

Catalog No. AAB2416903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 198206-33-6
    Molecular Formula C7H4F3IO
    Formula Weight 288.01
    MDL Number MFCD01090992
    Synonym 3-(Trifluoromethoxy)iodobenzene
    Chemical Name or Material 1-Iodo-3-(trifluoromethoxy)benzene

    Specifications

    Formula Weight 288.01
    Molecular Formula C7H4F3IO
    MDL Number MFCD01090992
    CAS 198206-33-6
    Boiling Point 185° to 186°C
    Synonym 3-(Trifluoromethoxy)iodobenzene
    Quantity 1g
    Flash Point 65°C (150°F)
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 1-Iodo-3-(trifluoromethoxy)benzene
    Assay ≥97%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit